Formamidine disulphide dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

供应商: TCI
Warning

Synonyms: Dithiourea Dihydrochloride, Dithiodiformamide Dihydrochloride

F0046-25G F0046-5G
TCIAF0046-25GEA 0 CNY
TCIAF0046-25G TCIAF0046-5G
Formamidine disulphide dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Formamidine disulphide dihydrochloride
分子式: C₂H₆N₄S₂
分子量: 223.13 g/mol
MDL编号: MFCD00012586
CAS号: 14807-75-1
EINECS: 238-876-3

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR