Ethyl-4-aminobutyrate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

供应商: TCI

Synonyms: Ethyl 4-aminobutanoate hydrochloride, Ethyl 4-Aminobutyrate HCl, 4-Aminobutyric Acid Ethyl Ester Hydrochloride, H-GABA-OEt.HCl

E0924-5G E0924-25G
TCIAE0924-5GEA 0 CNY
TCIAE0924-5G TCIAE0924-25G
Ethyl-4-aminobutyrate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Ethyl-4-aminobutyrate hydrochloride
分子式: C₆H₁₃NO₂
分子量: 167.63 g/mol
MDL编号: MFCD00012915
CAS号: 6937-16-2
EINECS: 230-063-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR