Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate ≥98.0% (by GC)
供应商: TCI
Synonyms:
Ethyl 1,4-Benzodioxane-2-carboxylate, Ethyl 2,3-dihydrobenzo[1,4]dioxine-2-carboxylate, 1,4-Benzodioxane-2-carboxylic acid ethyl ester
E0902-25G
E0902-5G
TCIAE0902-25GEA
0
CNY
TCIAE0902-25G
TCIAE0902-5G
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate ≥98.0% (by GC)
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate
分子式:
C₁₁H₁₂O₄ 分子量: 208.2100 g/mol 沸点: 115°C |
MDL编号:
MFCD01099774 CAS号: 4739-94-0 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...