Etomidate ≥98.0% (通过GC)

供应商: TCI
Warning

E0897-100MG E0897-500MG
TCIAE0897-100MGEA 0 CNY
TCIAE0897-100MG TCIAE0897-500MG
Etomidate ≥98.0% (通过GC)
Etomidate
分子式: C₁₄H₁₆N₂O₂
存储条件: Refrigerator
MDL编号: MFCD00869295
CAS号: 33125-97-2
EINECS: 251-385-9
UN号: 3077
ADR: 9,III
Merck Index: 14,03881

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C
Specific rotation [a]20/D +67.0 to +71.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR