5-Ethylpyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-Ethylquinolinic Acid

E0887-25G E0887-5G
TCIAE0887-25GEA 0 CNY
TCIAE0887-25G TCIAE0887-5G
5-Ethylpyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
5-Ethylpyridine-2,3-dicarboxylic acid
分子式: C₉H₉NO₄
分子量: 195.17 g/mol
溶点: 156°C
MDL编号: MFCD00071733
CAS号: 102268-15-5

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR