4-吡唑甲酸乙酯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: ethyl 1H-pyrazole-4-carboxylate, Ethyl 4-pyrazolecarboxylate

E0871-1G E0871-5G
TCIAE0871-1GEA 0 CNY
TCIAE0871-1G TCIAE0871-5G
4-吡唑甲酸乙酯 ≥98.0% (by GC)
4-吡唑甲酸乙酯
分子式: C₆H₈N₂O₂
分子量: 140.14 g/mol
沸点: 140°C
MDL编号: MFCD00010844
CAS号: 37622-90-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR