2,3-丁二烯酸乙酯 ≥95.0% (通过GC)

供应商: TCI
Warning

E0820-1G E0820-5G
TCIAE0820-1GEA 0 CNY
TCIAE0820-1G TCIAE0820-5G
2,3-丁二烯酸乙酯 ≥95.0% (通过GC)
分子式: C₆H₈O₂
沸点: 80°C
闪点: 47°C
存储条件: Freezer
MDL编号: MFCD01863567
CAS号: 14369-81-4
UN号: 3272
ADR: 3,III

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR