Ethyl-DL-leucate ≥98.0% (通过GC)
供应商: TCI
E0802-25G
E0802-5G
TCIAE0802-25GEA
0
CNY
TCIAE0802-25G
TCIAE0802-5G
Ethyl-DL-leucate ≥98.0% (通过GC)
Ethyl-DL-leucate
分子式:
C₈H₁₆O₃ 沸点: 185°C 存储条件: Refrigerator |
CAS号:
10348-47-7 EINECS: 233-760-9 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...