Epinastine hydrochloride ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
Synonyms:
3-Amino-9,13b-dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepine Hydrochloride
E0799-1G
E0799-100MG
TCIAE0799-1GEA
0
CNY
TCIAE0799-1G
TCIAE0799-100MG
Epinastine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Epinastine hydrochloride
分子式:
C₁₆H₁₅N₃ 分子量: 285.78 g/mol 溶点: 275°C 存储条件: Refrigerator |
MDL编号:
MFCD00933434 CAS号: 108929-04-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...