3,4-Epoxytetrahydrofuran ≥97.0% (通过GC)

供应商: TCI
Warning

E0795-25G E0795-5G
TCIAE0795-25GEA 0 CNY
TCIAE0795-25G TCIAE0795-5G
3,4-Epoxytetrahydrofuran ≥97.0% (通过GC)
3,4-Epoxytetrahydrofuran
分子式: C₄H₆O₂
沸点: 144°C
MDL编号: MFCD00800639
CAS号: 285-69-8
EINECS: 206-006-1
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR