3-溴-2-(溴甲基)丙酸乙酯 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Ethyl 3-bromo-2-(bromomethyl)propanoate, 3-Bromo-2-(bromomethyl)propionic Acid Ethyl Ester
E0791-25G
E0791-5G
TCIAE0791-25GEA
0
CNY
TCIAE0791-25G
TCIAE0791-5G
3-溴-2-(溴甲基)丙酸乙酯 ≥97.0% (by GC)
3-溴-2-(溴甲基)丙酸乙酯
分子式:
C₆H₁₀Br₂O₂ 分子量: 273.95 g/mol 沸点: 86°C |
MDL编号:
MFCD00034154 CAS号: 58539-11-0 |
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...