Ethyl 3-(Morpholino)propionate ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-[2-(Ethoxycarbonyl)ethyl]morpholine, Ethyl 3-morpholinopropanoate, 3-(Morpholino)propionic Acid Ethyl Ester

E0784-1G E0784-5G
TCIAE0784-1GEA 0 CNY
TCIAE0784-1G TCIAE0784-5G
Ethyl 3-(Morpholino)propionate ≥97.0% (by GC, titration analysis)
Ethyl-3-(morpholino)propionate
分子式: C₉H₁₇NO₃
沸点: 118°C
MDL编号: MFCD00014622
CAS号: 20120-24-5
EINECS: 243-526-8

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR