Ethionamide ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-Ethylpyridine-4-carbothioamide, 2-Ethylthioisonicotinamide
E0695-25G
E0695-5G
TCIAE0695-25GEA
0
CNY
TCIAE0695-25G
TCIAE0695-5G
Ethionamide ≥98.0% (by titrimetric analysis)
Ethionamide
分子式:
C₈H₁₀N₂S 分子量: 166.24 g/mol 沸点: 167°C |
MDL编号:
MFCD00057361 CAS号: 536-33-4 EINECS: 208-628-9 Merck Index: 14,03737 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...