Ethyl-4-Hydroxy-1-piperidinecarboxylate ≥98.0% (by GC)
供应商: TCI
Synonyms:
Ethyl 4-hydroxypiperidine-1-carboxylate, 1-Carbethoxy-4-hydroxypiperidine, 4-Hydroxy-1-piperidinecarboxylic Acid Ethyl Ester
E0656-25G
E0656-5G
TCIAE0656-25GEA
0
CNY
TCIAE0656-25G
TCIAE0656-5G
Ethyl-4-Hydroxy-1-piperidinecarboxylate ≥98.0% (by GC)
Ethyl-4-Hydroxy-1-piperidinecarboxylate
分子式:
C₈H₁₅NO₃ 分子量: 173.21 g/mol |
MDL编号:
MFCD00086880 CAS号: 65214-82-6 EINECS: 265-636-5 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...