乙酰乙酸甲基丙烯酸乙二醇酯 ≥95.0% (by HPLC) stabilised
供应商: TCI
|
Warning
|
Synonyms:
3-氧代丁酸-2-[2-甲基-2-丙烯酰基)氧]乙基酯, AAEM, Lonzamon AAMEA PQ, 2-甲基丙烯酸乙酰乙酸氧乙酯
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
E0489-25G
E0489-500G
TCIAE0489-25GEA
0
CNY
TCIAE0489-25G
TCIAE0489-500G
乙酰乙酸甲基丙烯酸乙二醇酯 ≥95.0% (by HPLC) stabilised
乙酰乙酸甲基丙烯酸乙二醇酯
分子式:
C₁₀H₁₄O₅ 分子量: 214.22 g/mol 闪点: 141°C |
MDL编号:
MFCD00054405 CAS号: 21282-97-3 EINECS: 244-311-1 |
产品技术规格
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (HPLC) | min. 95.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...