(R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide

供应商: TCI
Warning

Synonyms: (R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide, (R)-1-Ethylpyrrolidine-2-carboxamide, N-Ethyl-D-prolinamide

E0468-1G E0468-5G
TCIAE0468-1GEA 0 CNY
TCIAE0468-1G TCIAE0468-5G
(R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide
(R)-(+)-1-Ethyl-2-pyrrolidinecarboxamide
分子式: C₇H₁₄N₂O
MDL编号: MFCD00671532
CAS号: 381670-32-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 110.0 to 113.0 deg-C
Specific rotation [a]20/D +114.0 to +120.0 deg (C=0.5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR