2-烯丙氧基乙醇 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 乙二醇单烯丙基醚

E0326-25ML E0326-500ML
TCIAE0326-25MLEA 0 CNY
TCIAE0326-25ML TCIAE0326-500ML
2-烯丙氧基乙醇 ≥98.0% (by GC)
2-烯丙氧基乙醇
分子式: C₅H₁₀O₂
分子量: 102.13 g/mol
沸点: 161°C
闪点: 60°C
MDL编号: MFCD00020607
CAS号: 111-45-5
EINECS: 203-871-7
UN号: 3271
ADR: 3 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR