3-(2-Ethylhexyloxy)propylamine ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
3-Aminopropyl 2-Ethylhexyl ether, n-Propanolamine 2-Ethylhexyl ether, 3-Aminopropyl Octyl ether, 3-((2-Ethylhexyl)oxy)propan-1-amine
E0295-25ML
E0295-500ML
TCIAE0295-25MLEA
0
CNY
TCIAE0295-25ML
TCIAE0295-500ML
3-(2-Ethylhexyloxy)propylamine ≥98.0% (by GC, titration analysis)
3-(2-Ethylhexyloxy)propylamine
分子式:
C₁₁H₂₅NO 沸点: 78°C 闪点: 103°C |
MDL编号:
MFCD00025619 CAS号: 5397-31-9 EINECS: 226-420-6 UN号: 2810 ADR: 6.1,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...