Ethyl phosphate (mono and di ester mixture)

供应商: TCI
Danger

Synonyms: Phosphoric Acid Ethyl Ester

E0236-25ML E0236-500ML
TCIAE0236-25MLEA 0 CNY
TCIAE0236-25ML TCIAE0236-500ML
Ethyl phosphate (mono and di ester mixture)
Ethyl phosphate (mono and di ester mixture)
闪点: 193°C
MDL编号: MFCD00136002
CAS号: 37203-76-2
EINECS: 253-391-7
UN号: 1760
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Assay of mono ester 35.0 to 47.0 %
Assay of Diester 53.0 to 65.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR