辛基缩水甘油醚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Ethylhexylglycidylether, (2-Ethylhexyloxy)-2,3-epoxypropane
E0229-25ML
E0229-500ML
TCIAE0229-25MLEA
0
CNY
TCIAE0229-25ML
TCIAE0229-500ML
辛基缩水甘油醚 ≥98.0% (by GC)
辛基缩水甘油醚
分子式:
C₁₁H₂₂O₂ 沸点: 61°C 闪点: 94°C |
MDL编号:
MFCD00005141 CAS号: 2461-15-6 EINECS: 219-553-6 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...