N-Ethyltoluenesulphonamide(o and p mixture) ≥98.0% (by total nitrogen basis)

供应商: TCI

E0187-25G E0187-500G
TCIAE0187-25GEA 0 CNY
TCIAE0187-25G TCIAE0187-500G
N-Ethyltoluenesulphonamide(o and p mixture) ≥98.0% (by total nitrogen basis)
N-Ethyltoluenesulphonamide(o and p mixture)
分子式: C₉H₁₃NO₂S
分子量: 199.27 g/mol
闪点: 193°C
MDL编号: MFCD00048511
CAS号: 76902-32-4

现在订购

产品技术规格

Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR