乙缩醛 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 乙缩醛, 1,1-二乙氧基乙烷, 乙醛缩二乙醇

102220010 102220050 102222500
ACRO102220010EA 0 CNY
ACRO102220010 ACRO102220050 ACRO102222500
乙缩醛 99%
乙缩醛
分子式: C₆H₁₄O₂
分子量: 118.18 g/mol
沸点: 102.7
溶点: –100 °C
密度: 0.825 g/cm³
闪点: –21 °C
存储条件: Refrigerator
MDL编号: MFCD00009243
CAS号: 105-57-7
EINECS: 203-310-6
UN号: 1088
ADR: 3 II
Merck Index: 13,00040

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3790 to 1.3850 (20°C, 589 nm)
Density (20°C) 0.822 to 0.829 kg/l
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR