二甲基乙二肟 99+% ACS
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2,3-丁二酮二肟, 丁二酮肟
408330250
408331000
408335000
ACRO408330250EA
0
CNY
ACRO408330250
ACRO408331000
ACRO408335000
二甲基乙二肟 99+% ACS
二甲基乙二肟
分子式:
CH₃C(=NOH)C(=NOH)CH₃ 分子量: 116.12 g/mol 沸点: 53 °C (8 mmHg) 溶点: 239…242 °C 存储条件: Ambient |
MDL编号:
MFCD00002117 CAS号: 95-45-4 EINECS: 202-420-1 UN号: 2926 ADR: 4.1 III Merck Index: 13,03272 |
产品技术规格
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 239 to 242 °C (Dec) |
Residue after ignition | ≤0.05 % |
Gravimetry | ≥99.0 % |
Suitability | (for Ni determination) Passes test |
Insoluble matter | ≤0.05 % (in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...