二甲基乙二肟 99+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2,3-丁二酮二肟, 丁二酮肟

172001000 172005000
ACRO172001000EA 0 CNY
ACRO172001000 ACRO172005000
二甲基乙二肟 99+%
二甲基乙二肟
分子式: CH₃C(=NOH)C(=NOH)CH₃
分子量: 116.12 g/mol
沸点: 53
溶点: 239…242 °C (dec.)
存储条件: Ambient
MDL编号: MFCD00002117
CAS号: 95-45-4
EINECS: 202-420-1
UN号: 2926
ADR: 4.1 III
Merck Index: 13,03272

现在订购

产品技术规格

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 239 to 242 °C (Dec)
Sulfated ash ≤0.1 %
Gravimetry ≥99.0 %
Insoluble matter ≤0.05 % (in alcohol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR