Domperidone ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: 5-Chloro-1-[1-[3-(2-oxo-1-benzimidazolyl)propyl]-4-piperidyl]benzimidazol-2-one

D4125-1G D4125-200MG
TCIAD4125-1GEA 0 CNY
TCIAD4125-1G TCIAD4125-200MG
Domperidone ≥98.0% (by HPLC, titration analysis)
Domperidone
分子式: C₂₂H₂₄ClN₅O₂
分子量: 425.92 g/mol
溶点: 243°C
MDL编号: MFCD00069256
CAS号: 57808-66-9
EINECS: 260-968-7
Merck Index: 14,03418

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR