Diethyl-3,3'-iminodipropionate ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
Diethyl 3,3'-azanediyldipropanoate, 3,3'-Iminodipropionic Acid Diethyl Ester
D4111-1G
D4111-5G
TCIAD4111-1GEA
0
CNY
TCIAD4111-1G
TCIAD4111-5G
Diethyl-3,3'-iminodipropionate ≥98.0% (by GC, titration analysis)
Diethyl-3,3'-iminodipropionate
分子式:
C₁₀H₁₉NO₄ 分子量: 217.27 g/mol 沸点: 139°C 存储条件: Refrigerator |
MDL编号:
MFCD14705114 CAS号: 3518-88-5 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...