3-二甲胺基丙烯醛 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
3-(Dimethylamino)acrylaldehyde, 3-(Dimethylamino)propenal, 3-(Dimethylamino)-2-propenal
D4092-25G
D4092-5G
TCIAD4092-25GEA
0
CNY
TCIAD4092-25G
TCIAD4092-5G
3-二甲胺基丙烯醛 ≥98.0% (by GC, titration analysis)
3-二甲胺基丙烯醛
分子式:
C₅H₉NO 分子量: 99.13 g/mol 沸点: 91°C |
MDL编号:
MFCD00006999 CAS号: 927-63-9 EINECS: 213-157-7 UN号: 3267 ADR: 8 II |
产品技术规格
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...