3,4-Diaminofurazan ≥98.0% (通过GC)

供应商: TCI
Warning

D4030-1G D4030-5G
TCIAD4030-1GEA 0 CNY
TCIAD4030-1G TCIAD4030-5G
3,4-Diaminofurazan ≥98.0% (通过GC)
3,4-Diaminofurazan
分子式: C₂H₄N₄O
MDL编号: MFCD00138084
CAS号: 17220-38-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 95.0 %
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR