1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one

D3949-1G D3949-5G
TCIAD3949-1GEA 0 CNY
TCIAD3949-1G TCIAD3949-5G
1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one ≥98.0% (by GC)
1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one
分子式: C₁₀H₁₂N₂O
分子量: 176.22 g/mol
溶点: 71°C
MDL编号: MFCD00115180
CAS号: 55314-16-4

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR