富马酸二异丁酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: Fumaric Acid Diisobutyl Ester

D3915-25G D3915-500G
TCIAD3915-25GEA 0 CNY
TCIAD3915-25G TCIAD3915-500G
富马酸二异丁酯 ≥98.0% (by GC)
富马酸二异丁酯
分子式: C₁₂H₂₀O₄
分子量: 228.29 g/mol
沸点: 122°C
MDL编号: MFCD00152430
CAS号: 7283-69-4
EINECS: 230-706-6

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR