2,3-二氢苯并呋喃-5-甲醛 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
5-Coumarancarboxaldehyde, 2,3-Dihydrobenzofuran-5-carbaldehyde, 2,3-Dihydrobenzo[b]furan-5-carboxaldehyde, 2,3-Dihydrobenzofuran-5-carboxaldehyde
D3857-25G
D3857-5G
TCIAD3857-25GEA
0
CNY
TCIAD3857-25G
TCIAD3857-5G
2,3-二氢苯并呋喃-5-甲醛 ≥96.0% (by GC)
2,3-二氢苯并呋喃-5-甲醛
分子式:
C₉H₈O₂ 分子量: 148.16 g/mol 沸点: 96°C |
MDL编号:
MFCD00068058 CAS号: 55745-70-5 EINECS: 259-788-1 |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...