4,4'-Di(1,2,3-triazolyl) Disulphide Hydrate ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 4,4'-Dithiobis(1,2,3-triazole)

D3855-1G D3855-5G
TCIAD3855-1GEA 0 CNY
TCIAD3855-1G TCIAD3855-5G
4,4'-Di(1,2,3-triazolyl) Disulphide Hydrate ≥98.0% (by HPLC, titration analysis)
4,4'-Di(1,2,3-triazolyl) Disulphide Hydrate
分子式: C₄H₄N₆S₂
分子量: 200.24 g/mol
CAS号: 6440-09-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR