3,5-Dibromodithieno[3,2-b:2',3'-d]thiophene ≥96.0% (通过GC)

供应商: TCI

D3800-1G D3800-200MG
TCIAD3800-1GEA 0 CNY
TCIAD3800-1G TCIAD3800-200MG
3,5-Dibromodithieno[3,2-b:2',3'-d]thiophene ≥96.0% (通过GC)
3,5-Dibromodithieno[3,2-b:2',3'-d]thiophene
分子式: C₈H₂Br₂S₃
CAS号: 502764-54-7

现在订购

产品技术规格

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 235.0 to 241.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR