Dibromoisocyanuric acid ≥97.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
DBI, 1,3-Dibromo-1,3,5-triazine-2,4,6-trione
D3753-25G
D3753-5G
TCIAD3753-25GEA
0
CNY
TCIAD3753-25G
TCIAD3753-5G
Dibromoisocyanuric acid ≥97.0% (by titrimetric analysis)
Dibromoisocyanuric acid
分子式:
C₃HBr₂N₃O₃ 溶点: 309°C 存储条件: Refrigerator |
MDL编号:
MFCD00463941 CAS号: 15114-43-9 UN号: 3085 ADR: 5.1,II |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...