3,6-Dichlorocarbazole ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: 3,6-Dichloro-9H-carbazole

D3751-1G D3751-5G
TCIAD3751-1GEA 0 CNY
TCIAD3751-1G TCIAD3751-5G
3,6-Dichlorocarbazole ≥96.0% (by GC)
3,6-Dichlorocarbazole
分子式: C₁₂H₇Cl₂N
MDL编号: MFCD00218271
CAS号: 5599-71-3

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 204.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR