N-甲氧基-N-甲羰基甲基膦酸二乙酯 ≥97.0% (by GC)

供应商: TCI

Synonyms: Diethyl N-methoxy-N-methylphosphonoacetamide, N-Methoxy-N-methyl-phosphonoacetamide diethyl ester

D3708-1G D3708-5G
TCIAD3708-1GEA 0 CNY
TCIAD3708-1G TCIAD3708-5G
N-甲氧基-N-甲羰基甲基膦酸二乙酯 ≥97.0% (by GC)
N-甲氧基-N-甲羰基甲基膦酸二乙酯
分子式: C₈H₁₈NO₅P
分子量: 239.21 g/mol
沸点: 124°C
MDL编号: MFCD00134233
CAS号: 124931-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d37022261b47865107db721e6d59efbe2ba14ce4&vid=d37022261b47865107db721e6d59efbe2ba14ce4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR