2,6-Dimethylbenzothiazole ≥98.0% (by GC)

供应商: TCI
Warning

D3674-25G D3674-5G
TCIAD3674-25GEA 0 CNY
TCIAD3674-25G TCIAD3674-5G
2,6-Dimethylbenzothiazole ≥98.0% (by GC)
2,6-Dimethylbenzothiazole
分子式: C₉H₉NS
沸点: 132°C
CAS号: 2941-71-1
EINECS: 220-930-2

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR