(+)-cis-Diltiazem hydrochloride ≥98.0% (通过HPLC,滴定分析)

供应商: TCI
Warning

D3662-25G D3662-5G
TCIAD3662-25GEA 0 CNY
TCIAD3662-25G TCIAD3662-5G
(+)-cis-Diltiazem hydrochloride ≥98.0% (通过HPLC,滴定分析)
(+)-cis-Diltiazem hydrochloride
分子式: C₂₂H₂₆N₂O₄S·HCl
溶点: 210°C
存储条件: Refrigerator
MDL编号: MFCD00069252
CAS号: 33286-22-5
EINECS: 251-443-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D 114 to 122 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR