2'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 5-Fluoro-2'-deoxycytidine, 4-Amino-5-fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

D3642-1G D3642-5G
TCIAD3642-1GEA 0 CNY
TCIAD3642-1G TCIAD3642-5G
2'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)
2'-Deoxy-5-fluorocytidine
分子式: C₉H₁₂FN₃O₄
分子量: 245.21 g/mol
溶点: 196°C
MDL编号: MFCD00077348
CAS号: 10356-76-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +65.0 to +75.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR