Diethyl-trans-1,4-cyclohexanedicarboxylate ≥98.0% (通过GC)

供应商: TCI

D3639-25G D3639-5G
TCIAD3639-25GEA 0 CNY
TCIAD3639-25G TCIAD3639-5G
Diethyl-trans-1,4-cyclohexanedicarboxylate ≥98.0% (通过GC)
Diethyl-trans-1,4-cyclohexanedicarboxylate
分子式: C₁₂H₂₀O₄
CAS号: 19145-96-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR