Dacarbazine ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 5- (3,3-Dimethyl-1-triazenyl) imidazole-4-carboxamide, DTIC, 5-(3,3-Dimethyltriazeno)imidazole-4-carboxamide

D3634-1G D3634-5G
TCIAD3634-1GEA 0 CNY
TCIAD3634-1G TCIAD3634-5G
Dacarbazine ≥98.0% (by HPLC, titration analysis)
Dacarbazine
分子式: C₆H₁₀N₆O
分子量: 182.19 g/mol
溶点: 255°C
存储条件: Refrigerator
MDL编号: MFCD00057167
CAS号: 4342-03-4
EINECS: 224-396-1
Merck Index: 14,02798

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR