Diethyl-2-ethyl-2-methylmalonate ≥97.0% (by GC)

供应商: TCI

Synonyms: 2-Ethyl-2-methylmalonic Acid Diethyl Ester

D3630-25G D3630-5G
TCIAD3630-25GEA 0 CNY
TCIAD3630-25G TCIAD3630-5G
Diethyl-2-ethyl-2-methylmalonate ≥97.0% (by GC)
Diethyl-2-ethyl-2-methylmalonate
分子式: C₁₀H₁₈O₄
分子量: 202.25 g/mol
沸点: 79°C
MDL编号: MFCD02258541
CAS号: 2049-70-9
EINECS: 218-070-8

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR