2'-Deoxy-5-methylcytidine ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine

D3610-100MG D3610-5G D3610-500MG
TCIAD3610-100MGEA 0 CNY
TCIAD3610-100MG TCIAD3610-5G TCIAD3610-500MG
2'-Deoxy-5-methylcytidine ≥98.0% (by HPLC, titration analysis)
2'-Deoxy-5-methylcytidine
分子式: C₁₀H₁₅N₃O₄
分子量: 241.25 g/mol
溶点: 193°C
MDL编号: MFCD00006549
CAS号: 838-07-3
EINECS: 212-655-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +42.0 to +46.0 deg (C=1.4, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR