Dimethyl octafluoroadipate ≥97.0% (by GC)

供应商: TCI

Synonyms: Dimethyl Octafluorohexanedioate, Dimethyl Perfluoroadipate, Octafluoroadipic Acid Dimethyl Ester, Perfluoroadipic Acid Dimethyl Ester, Octafluorohexanedioic Acid Dimethyl Ester

D3590-1G D3590-5G
TCIAD3590-1GEA 0 CNY
TCIAD3590-1G TCIAD3590-5G
Dimethyl octafluoroadipate ≥97.0% (by GC)
Dimethyl octafluoroadipate
分子式: C₈H₆F₈O₄
沸点: 123°C
MDL编号: MFCD00042267
CAS号: 3107-98-0

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR