5′-O-(4,4′-二甲氧基三苯甲基)胸苷 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: DMT-T

D3566-25G D3566-5G
TCIAD3566-25GEA 0 CNY
TCIAD3566-25G TCIAD3566-5G
5′-O-(4,4′-二甲氧基三苯甲基)胸苷 ≥98.0% (by HPLC, titration analysis)
5′-O-(4,4′-二甲氧基三苯甲基)胸苷
分子式: C₃₁H₃₂N₂O₇
分子量: 544.60 g/mol
存储条件: Refrigerator
MDL编号: MFCD00010113
CAS号: 40615-39-2
EINECS: 255-003-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +5.0 to +9.0 deg (C=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR