Dimethyl hexadecanedioate ≥95.0% (by GC)

供应商: TCI

Synonyms: Hexadecanedioic Acid Dimethyl Ester

D3554-25G D3554-5G
TCIAD3554-25GEA 0 CNY
TCIAD3554-25G TCIAD3554-5G
Dimethyl hexadecanedioate ≥95.0% (by GC)
Dimethyl hexadecanedioate
分子式: C₁₈H₃₄O₄
沸点: 172°C
MDL编号: MFCD00039806
CAS号: 19102-90-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 50.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR