[N-[ɑ-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel ≥95.0% (通过滴定分析)
供应商: TCI
|
Danger
|
D3543-1G
D3543-100MG
TCIAD3543-1GEA
0
CNY
TCIAD3543-1G
TCIAD3543-100MG
[N-[ɑ-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel ≥95.0% (通过滴定分析)
[N-[alpha-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel
分子式:
C₂₅H₃₁N₃NiO₃ |
CAS号:
847654-16-4 |
产品技术规格
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...