1,3-Di-O-benzyl-2-O-lauroylglycerol ≥85.0% (by GC)
供应商: TCI
Synonyms:
1,3-Bis(benzyloxy)isopropyl Dodecanoate, Glycerol 1,3-Dibenzyl Ether 2-Laurate
D3524-1G
D3524-5G
TCIAD3524-1GEA
0
CNY
TCIAD3524-1G
TCIAD3524-5G
1,3-Di-O-benzyl-2-O-lauroylglycerol ≥85.0% (by GC)
1,3-Di-O-benzyl-2-O-lauroylglycerol
分子式:
C₂₉H₄₂O₄ 分子量: 454.65 g/mol |
CAS号:
519177-07-2 |
产品技术规格
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 85.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...