2-二环己膦基-2′-(N,N-二甲胺)联苯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: DavePhos

D3389-1G D3389-5G
TCIAD3389-1GEA 0 CNY
TCIAD3389-1G TCIAD3389-5G
2-二环己膦基-2′-(N,N-二甲胺)联苯 ≥98.0% (by GC, titration analysis)
2-二环己膦基-2′-(N,N-二甲胺)联苯
分子式: C₂₆H₃₆NP
分子量: 393.55 g/mol
MDL编号: MFCD02183572
CAS号: 213697-53-1

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR