Dicyclopentanyl methacrylate ≥95.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: Methacrylic Acid Tetrahydrodicyclopentadienyl Ester, Octahydro-1H-4,7-methanoinden-5-yl methacrylate, Methacrylic Acid Dicyclopentanyl Ester, Tetrahydrodicyclopentadienyl Methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D3381-25G D3381-500G
TCIAD3381-25GEA 0 CNY
TCIAD3381-25G TCIAD3381-500G
Dicyclopentanyl methacrylate ≥95.0% (by GC) stabilised
Dicyclopentanyl methacrylate
分子式: C₁₄H₂₀O₂
闪点: 145°C
存储条件: Freezer
MDL编号: MFCD28963492
CAS号: 34759-34-7

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR